Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Septa, red PTFE/Silicone
Products
Product Category
septa (3)
Brand
Supelco (3)
Diameter (mm)
OD (mm)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
27097-U
Septa, red PTFE/Silicone,
red PTFE/white silicone, diam. × thickness 8 mm × 1.5 mm, pkg of 100 ea
Expand
View Pricing
29039-U
Septa, red PTFE/Silicone,
red PTFE/silicone, diam. × thickness 9 mm × 1.0 mm, pkg of 100 ea
Expand
View Pricing
27277
Septa, red PTFE/Silicone,
red PTFE/white silicone, diam. × thickness 10 mm × 0.060 in., for use with 2 mL, 10/425 thread, large opening, pkg of 100 ea
Expand
View Pricing