Skip to Content
Products
Cart
0
AG
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Back
Septa, PTFE/Silicone
Products
Product Category
septa (7)
Brand
Supelco (7)
Manufacturer
Diameter (mm)
OD (mm)
Color
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
27236
Septa, PTFE/Silicone,
tan PTFE/silicone septa, diam. × thickness × PTFE thickness 20 mm × 0.125 in. × 2 mil, pkg of 100 ea
Expand
View Pricing
507482
Septa, PTFE/Silicone,
red PTFE/white silicone, diam. 12 mm, pkg of 100 ea
Expand
View Pricing
27539
Septa, PTFE/Silicone,
blue PTFE/white silicone, diam. × thickness 20 mm × 0.75 mm, pkg of 100 ea
Expand
View Pricing
23242-U
Septa, PTFE/Silicone,
tan PTFE/silicone septum, septum diam. × total thickness × PTFE thickness 18 mm × 0.060 in. × 2 mil, pkg of 144 ea
Expand
View Pricing
23245-U
Septa, PTFE/Silicone,
PTFE/silicone, septum diam. × thickness 22 mm × 0.100 in., pkg of 144 ea
Expand
View Pricing
27237-U
Septa, PTFE/Silicone,
transparent PTFE/silicone, diam. × thickness × PTFE thickness 20 mm × 0.100 in. × 10 mil, pkg of 100 ea
Expand
View Pricing
508616
Septa, PTFE/Silicone,
blue PTFE/silicone, diam. × thickness 20 mm × 0.045 in., hardness 30 (Durometer: shore A), pkg of 100 ea
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
English - EN
Learn More