Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Self standing screw cap tubes and attached caps with O-rings
Products
Product Category
tubes (17)
Manufacturer
Capacity (mL)
Color
Package Size
Sterility
Filter Products
Showing 1-17 of 17
Compare
Product Number
Description
Pricing
AXYST150SSCS
Self standing screw cap tubes and attached caps with O-rings,
capacity 1.5 mL, clear, sterile
Expand
View Pricing
AXYSCT050SSBS
Self standing screw cap tubes and attached caps with O-rings,
capacity 0.5 mL, blue, sterile
Expand
View Pricing
AXYSCT200SSGS
Self standing screw cap tubes and attached caps with O-rings,
capacity 2.0 mL, green, sterile
Expand
View Pricing
AXYSCT150SSBS
Self standing screw cap tubes and attached caps with O-rings,
capacity 0.5 mL, blue, sterile
Expand
View Pricing
AXYSCT200SSBS
Self standing screw cap tubes and attached caps with O-rings,
capacity 2.0 mL, blue, sterile
Expand
View Pricing
AXYSCT150SSX
Self standing screw cap tubes and attached caps with O-rings,
capacity 0.5 mL, amber, with brown caps, sterile
Expand
View Pricing
AXYSCT45SSCS
Self standing screw cap tubes and attached caps with O-rings,
capacity 0.6 mL, clear, sterile
Expand
View Pricing
AXYSCT200SSOS
Self standing screw cap tubes and attached caps with O-rings,
capacity 2.0 mL, orange, sterile
Expand
View Pricing
AXYSCT200SSRS
Self standing screw cap tubes and attached caps with O-rings,
capacity 2.0 mL, red, sterile
Expand
View Pricing
AXYSCT150SSRS
Self standing screw cap tubes and attached caps with O-rings,
capacity 0.5 mL, red, sterile
Expand
View Pricing
AXYSCT200SSYS
Self standing screw cap tubes and attached caps with O-rings,
capacity 2.0 mL, yellow, sterile
Expand
View Pricing
AXYSCT050SSOS
Self standing screw cap tubes and attached caps with O-rings,
capacity 0.5 mL, orange, sterile
Expand
View Pricing
AXYSCT050SSWS
Self standing screw cap tubes and attached caps with O-rings,
capacity 0.5 mL, white, sterile
Expand
View Pricing
AXYSCT050SSYS
Self standing screw cap tubes and attached caps with O-rings,
capacity 0.5 mL, yellow, sterile
Expand
View Pricing
AXYSCT150SSGS
Self standing screw cap tubes and attached caps with O-rings,
capacity 0.5 mL, green, sterile
Expand
View Pricing
AXYSCT050SSRS
Self standing screw cap tubes and attached caps with O-rings,
capacity 0.5 mL, red, sterile
Expand
View Pricing
AXYSCT050SSGS
Self standing screw cap tubes and attached caps with O-rings,
capacity 0.5 mL, green, sterile
Expand
View Pricing