Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ruthenium on carbon
Empirical Formula (Hill Notation):
Ru
Molecular Weight:
101.07
Products
Brand
Sigma-Aldrich (2)
Sustainability
Feature
Formula Weight
Physical Form
Reagent Type
Core
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
206180
Ruthenium on carbon,
extent of labeling: 5 wt. % loading
Expand
View Pricing
908045
Ruthenium on carbon,
Evonik Noblyst
®
P3059 5% Ru
Expand
View Pricing
907871
Ruthenium on carbon,
Evonik Noblyst
®
P3060 5% Ru
Expand
View Pricing