Skip to Content
Merck
Ruthenium Ruthenium, pellets, 2.5g, max. size 10mm, 99.9%

Ruthenium

Ruthenium Ruthenium, pellets, 2.5g, max. size 10mm, 99.9%
Synonym(s):
Ruthenium
Empirical Formula (Hill Notation):
Ru
CAS Number:
Molecular Weight:
101.07

Products

Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Length (mm)
Element
Quality Segment
Particle Size (µm)
Showing 1-17 of 17
Compare
Product Number
Description
Pricing