Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Reagent Alcohol
Synonym(s):
Ethanol, RGA, denatured alcohol
CAS Number:
64-17-5
Products
Product Category
solvents (8)
Brand
Sigma-Aldrich (8)
Biological Source
Color
Physical Form
Quality Segment
Application
Special Grade
Anhydrous (2)
Technique
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
270741
Reagent Alcohol,
≥89.0% (GC), denatured, suitable for HPLC, contains 5% isopropyl alcohol as denaturant, 5% methyl alcohol as denaturant
Expand
View Pricing
362808
Reagent Alcohol,
≥89% (GC), reagent grade, contains 5% isopropyl alcohol and 5% methyl alcohol as denaturant
Expand
View Pricing
R3154
Reagent Alcohol,
70%, used for histology tissue preparation
Expand
View Pricing
R8382
Reagent Alcohol,
used for histology tissue preparation
Expand
View Pricing
277649
Reagent Alcohol,
anhydrous reagent, contains 5% isopropyl alcohol and 5% methyl alcohol as denaturant
Expand
View Pricing
R3404
Reagent Alcohol,
95%, Used for histology tissue preparation
Expand
View Pricing
676829
Reagent Alcohol,
anhydrous, contains 5% isopropyl alcohol and 5% methyl alcohol as denaturant
Expand
View Pricing
R3279
Reagent Alcohol,
80%, used for histology tissue preparation
Expand
View Pricing