
Ranolazine

Synonym(s):
(±)-4-[2-Hydroxy-3-(o-methoxyphenoxy)propyl]-1-piperazineaceto-2′,6′-xylidide, (±)-N-(2,6-Dimethylphenyl)-4-[2-hydroxy-3-(2-methoxyphenoxy)propyl]piperazin-1-ylacetamide
Empirical Formula (Hill Notation):
C24H33N3O4
CAS Number:
Molecular Weight:
427.54
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade