Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
radiello
™
Diffusive Bodies
Products
Color
Manufacturer
Quality Segment
Application
Package Size
Pore size (µm)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
RAD120
radiello
™
Diffusive Bodies,
white, configured for general use, pk of 20
Expand
View Pricing
RAD1202
radiello
™
Diffusive Bodies,
yellow, configured for reduced sampling rates, pk of 20
Expand
View Pricing
RAD1201
radiello
™
Diffusive Bodies,
blue, configured for sampling light sensitive compounds, pk of 20
Expand
View Pricing
RAD1203
radiello
™
Diffusive Bodies,
permeative, configured for anaesthetic gases and vapors, pk of 20
Expand
View Pricing