Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Quartz - fused
Products
Manufacturer
Physical Form
Length (mm)
OD (mm)
Filter Products
Showing 1-20 of 119
Page 1 of 6
Page 1 of 6
Compare
Product Number
Description
Pricing
GF72061354
Quartz - fused,
sheet, thickness 0.25 mm, size 25 × 25 mm, condition polished both sides
Expand
View Pricing
GF72169533
Quartz - fused,
sheet, thickness 3.0 mm, size 34 × 89 mm, condition polished both sides
Expand
View Pricing
GF44607191
Quartz - fused,
tube, outside diameter 40 mm, length 200 mm, wall thickness 2 mm
Expand
View Pricing
GF03368819
Quartz - fused,
sheet, thickness 1.0 mm, size 50 × 50 mm, condition polished both sides
Expand
View Pricing
GF13892834
Quartz - fused,
powder, max. particle size 45 micron, weight 1000 g
Expand
View Pricing
GF58607950
Quartz - fused,
tube, outside diameter 1.0 mm, length 60 mm, wall thickness 0.25 mm
Expand
View Pricing
GF16318087
Quartz - fused,
sheet, thickness 0.50 mm, size 10 × 10 mm, condition polished both sides
Expand
View Pricing
GF73626958
Quartz - fused,
sheet, thickness 1.0 mm, size 25 × 25 mm, condition polished both sides
Expand
View Pricing
GF43962936
Quartz - fused,
powder, max. particle size 45 micron, weight 100 g
Expand
View Pricing
GF56120554
Quartz - fused,
tube, outside diameter 25 mm, length 100 mm, wall thickness 1.5 mm
Expand
View Pricing
GF57321277
Quartz - fused,
granule, 3 mm nominal granule size, weight 50 g, purity 99.999%
Expand
View Pricing
GF01488876
Quartz - fused,
tube, outside diameter 40 mm, length 100 mm, wall thickness 2 mm
Expand
View Pricing
GF60178350
Quartz - fused,
tube, outside diameter 5.0 mm, length 1000 mm, wall thickness 1.0 mm
Expand
View Pricing
GF56306965
Quartz - fused,
rod, 1.0 mm diameter, length 100 mm
Expand
View Pricing
GF24688143
Quartz - fused,
tube, outside diameter 1.0 mm, length 100 mm, wall thickness 0.1 mm
Expand
View Pricing
GF20911148
Quartz - fused,
powder, max. particle size 45 micron, weight 500 g
Expand
View Pricing
GF91178297
Quartz - fused,
tube, outside diameter 0.7 mm, length 100 mm, wall thickness 0.1 mm
Expand
View Pricing
GF41644874
Quartz - fused,
granule, 2-3 mm nominal granule size, weight 50 g
Expand
View Pricing
GF32301997
Quartz - fused,
granule, 0.85-1.7 mm nominal granule size, weight 50 g
Expand
View Pricing
GF91214423
Quartz - fused,
tube, outside diameter 40 mm, length 1000 mm, wall thickness 2 mm
Expand
View Pricing
Page 1 of 6
Page 1 of 6