Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pyrex
®
watch glasses
Synonym(s):
Corning
®
9985
Products
Manufacturer
OD (mm)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
CLS9985100
Pyrex
®
watch glasses,
O.D. 100 mm
Expand
View Pricing
CLS9985125
Pyrex
®
watch glasses,
O.D. 125 mm
Expand
View Pricing
CLS998590
Pyrex
®
watch glasses,
O.D. 90 mm
Expand
View Pricing
CLS9985150
Pyrex
®
watch glasses,
O.D. 150 mm
Expand
View Pricing
CLS998565
Pyrex
®
watch glasses,
O.D. 65 mm
Expand
View Pricing
CLS998575
Pyrex
®
watch glasses,
O.D. 75 mm
Expand
View Pricing