Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pyrex
®
volumetric pipette, to contain, to deliver, Class A
Synonym(s):
Corning
®
7103
Products
Product Category
pipettes (11)
Color
Manufacturer
Capacity (mL)
Length (mm)
Glassware Class
Package Size
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
CLS71036
Pyrex
®
volumetric pipette, to contain, to deliver, Class A,
orange, volume 6 mL, accuracy: 0.01 mL
Expand
View Pricing
CLS710310
Pyrex
®
volumetric pipette, to contain, to deliver, Class A,
red, volume 10 mL, accuracy: 0.02 mL
Expand
View Pricing
CLS71038
Pyrex
®
volumetric pipette, to contain, to deliver, Class A,
blue, volume 8 mL, accuracy: 0.02 mL
Expand
View Pricing
CLS71033
Pyrex
®
volumetric pipette, to contain, to deliver, Class A,
black, volume 3 mL, accuracy: 0.01 mL
Expand
View Pricing
CLS71037
Pyrex
®
volumetric pipette, to contain, to deliver, Class A,
green, volume 7 mL, accuracy: 0.01 mL
Expand
View Pricing
CLS71034
Pyrex
®
volumetric pipette, to contain, to deliver, Class A,
red, volume 4 mL, accuracy: 0.01 mL
Expand
View Pricing
CLS710315
Pyrex
®
volumetric pipette, to contain, to deliver, Class A,
green, volume 15 mL, accuracy: 0.03 mL
Expand
View Pricing
CLS710320
Pyrex
®
volumetric pipette, to contain, to deliver, Class A,
yellow, volume 20 mL, accuracy: 0.03 mL
Expand
View Pricing
CLS71031
Pyrex
®
volumetric pipette, to contain, to deliver, Class A,
blue, volume 1 mL, accuracy: 0.006 mL
Expand
View Pricing
CLS71032
Pyrex
®
volumetric pipette, to contain, to deliver, Class A,
orange, volume 2 mL, accuracy: 0.006 mL
Expand
View Pricing
CLS710325
Pyrex
®
volumetric pipette, to contain, to deliver, Class A,
blue, volume 25 mL, accuracy: 0.03 mL
Expand
View Pricing