Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pyrex
®
VISTA
™
volumetric pipette, Class A
Synonym(s):
Corning
®
70710
Products
Product Category
pipettes (4)
Manufacturer
Capacity (mL)
Glassware Class
Package Size
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
CLS707101
Pyrex
®
VISTA
™
volumetric pipette, Class A,
volume 1 mL
Expand
View Pricing
CLS7071010
Pyrex
®
VISTA
™
volumetric pipette, Class A,
volume 10 mL
Expand
View Pricing
CLS707105
Pyrex
®
VISTA
™
volumetric pipette, Class A,
volume 5 mL
Expand
View Pricing
CLS707102
Pyrex
®
VISTA
™
volumetric pipette, Class A,
volume 2 mL
Expand
View Pricing