Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pyrex
®
round-bottom boiling flask, short neck
Products
Product Category
flasks (21)
Manufacturer
Capacity (mL)
Closure Type
Package Size
Sterility
Style
Filter Products
Showing 1-20 of 21
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
CLS43205L
Pyrex
®
round-bottom boiling flask, short neck,
capacity 5,000 mL
Expand
View Pricing
CLS43211L
Pyrex
®
round-bottom boiling flask, short neck,
capacity 1,000 mL, joint: ST/NS 29/42
Expand
View Pricing
CLS4321A25
Pyrex
®
round-bottom boiling flask, short neck,
capacity 25 mL, joint: ST/NS 14/20
Expand
View Pricing
CLS43201L
Pyrex
®
round-bottom boiling flask, short neck,
capacity 1,000 mL
Expand
View Pricing
CLS4321A50
Pyrex
®
round-bottom boiling flask, short neck,
capacity 50 mL, joint: ST/NS 14/20
Expand
View Pricing
CLS4320100
Pyrex
®
round-bottom boiling flask, short neck,
capacity 100 mL
Expand
View Pricing
CLS4320250
Pyrex
®
round-bottom boiling flask, short neck,
capacity 250 mL
Expand
View Pricing
CLS4321A100
Pyrex
®
round-bottom boiling flask, short neck,
capacity 100 mL, joint: ST/NS 14/20
Expand
View Pricing
CLS4320500
Pyrex
®
round-bottom boiling flask, short neck,
capacity 500 mL
Expand
View Pricing
CLS4320A50
Pyrex
®
round-bottom boiling flask, short neck,
capacity 50 mL, joint: ST/NS 24/40
Expand
View Pricing
CLS43202L
Pyrex
®
round-bottom boiling flask, short neck,
capacity 2,000 mL
Expand
View Pricing
CLS4321250
Pyrex
®
round-bottom boiling flask, short neck,
capacity 250 mL, joint: ST/NS 29/42
Expand
View Pricing
CLS4320B250
Pyrex
®
round-bottom boiling flask, short neck,
capacity 250 mL, joint: ST/NS 19/22
Expand
View Pricing
CLS4320B100
Pyrex
®
round-bottom boiling flask, short neck,
capacity 100 mL, joint: ST/NS 19/22
Expand
View Pricing
CLS4321100
Pyrex
®
round-bottom boiling flask, short neck,
capacity 100 mL, joint: ST/NS 29/42
Expand
View Pricing
CLS4320B25
Pyrex
®
round-bottom boiling flask, short neck,
capacity 25 mL, joint: ST/NS 19/22
Expand
View Pricing
CLS4320B500
Pyrex
®
round-bottom boiling flask, short neck,
capacity 500 mL, joint: ST/NS 19/22
Expand
View Pricing
CLS4320B50
Pyrex
®
round-bottom boiling flask, short neck,
capacity 50 mL, joint: ST/NS 19/22
Expand
View Pricing
CLS4321500
Pyrex
®
round-bottom boiling flask, short neck,
capacity 500 mL, joint: ST/NS 29/42
Expand
View Pricing
CLS4321A250
Pyrex
®
round-bottom boiling flask, short neck,
capacity 250 mL, joint: ST/NS 14/20
Expand
View Pricing
Page 1 of 2
Page 1 of 2