Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
PYREXPLUS
®
coated solution bottles
Synonym(s):
Corning
®
61596
Products
Product Category
bottles (3)
Manufacturer
Capacity (L)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
CLS615969L
PYREXPLUS
®
coated solution bottles,
capacity 9.5 L
Expand
View Pricing
CLS6159613L
PYREXPLUS
®
coated solution bottles,
capacity 13.25 L
Expand
View Pricing
CLS6159619L
PYREXPLUS
®
coated solution bottles,
capacity 19.0 L
Expand
View Pricing