Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pyrex
®
Milk Dilution bottle with screw cap
Products
Product Category
bottles (4)
Manufacturer
Capacity (mL)
Diameter (mm)
Cap
Bottle material
Color
Vial Bottle Material
Vial Bottle Color
Package Size
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
CLS70368160
Pyrex
®
Milk Dilution bottle with screw cap,
capacity 160 mL, wide mouth
Expand
View Pricing
CLS70367160
Pyrex
®
Milk Dilution bottle with screw cap,
capacity 160 mL, narrow mouth
Expand
View Pricing
CLS70373160
Pyrex
®
Milk Dilution bottle with screw cap,
capacity 160 mL, wide mouth, graduated
Expand
View Pricing
CLS70372160
Pyrex
®
Milk Dilution bottle with screw cap,
capacity 160 mL, narrow mouth, graduated
Expand
View Pricing