Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pyrex
®
glass drying trays
Synonym(s):
Corning
®
3175
Products
Product Category
desiccators (3)
Manufacturer
Capacity (L)
Length (in)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
CLS31758
Pyrex
®
glass drying trays,
capacity 2 L, L × W × H 8 in. × 8 in. × 2 in.
Expand
View Pricing
CLS317510
Pyrex
®
glass drying trays,
capacity 3.5 L, L × W × H 15 in. × 10 in. × 2 in.
Expand
View Pricing
CLS31757
Pyrex
®
glass drying trays,
capacity 1.75 L, L × W × H 11 in. × 7 in. × 1 1/2 in.
Expand
View Pricing