Skip to Content
Merck
p-Xylene SAJ first grade, ≥99.0%

p-Xylene

p-Xylene SAJ first grade, ≥99.0%
Synonym(s):
1,4-Dimethylbenzene, p-Xylene
Linear Formula:
C6H4(CH3)2
CAS Number:
Molecular Weight:
106.17

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique