Skip to Content
Merck
Pullulan suitable for substrate for pullulanase

Pullulan

Pullulan suitable for substrate for pullulanase
CAS Number:
EC Number:

Products

Product Category
Brand
Color
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing