Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
PTFE stirrer guides
Products
Manufacturer
Diameter (mm)
Filter Products
Showing 1-13 of 13
Compare
Product Number
Description
Pricing
Z649317
PTFE stirrer guides,
shaft D 10 mm, joint: ST/NS 24/40
Expand
View Pricing
Z649422
PTFE stirrer guides,
shaft D 8 mm, joint: ST/NS 24/29
Expand
View Pricing
Z649287
PTFE stirrer guides,
shaft D 6 mm, joint: ST/NS 24/40
Expand
View Pricing
Z649430
PTFE stirrer guides,
shaft D 10 mm, joint: ST/NS 24/29
Expand
View Pricing
Z649309
PTFE stirrer guides,
shaft D 8 mm, joint: ST/NS 24/40
Expand
View Pricing
Z649414
PTFE stirrer guides,
shaft D 8 mm, joint: ST/NS 19/26
Expand
View Pricing
Z649325
PTFE stirrer guides,
shaft D 10 mm, joint: ST/NS 29/42
Expand
View Pricing
Z649406
PTFE stirrer guides,
shaft D 6 mm, joint: ST/NS 24/29
Expand
View Pricing
Z649481
PTFE stirrer guides,
shaft D 16 mm, joint: ST/NS 34/35
Expand
View Pricing
Z649465
PTFE stirrer guides,
shaft D 12 mm, joint: ST/NS 29/32
Expand
View Pricing
Z649457
PTFE stirrer guides,
shaft D 10 mm, joint: ST/NS 34/35
Expand
View Pricing
Z649341
PTFE stirrer guides,
shaft D 12 mm, joint: ST/NS 29/42
Expand
View Pricing
Z649392
PTFE stirrer guides,
shaft D 6 mm, joint: ST/NS 19/26
Expand
View Pricing