Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
p
-Rosolic acid
Synonym(s):
4-[Bis(4-hydroxyphenyl)methylene]-2,5-cyclohexadienone, Aurin
Empirical Formula (Hill Notation):
C
19
H
14
O
3
CAS Number:
603-45-2
Molecular Weight:
290.31
EC Number:
210-041-8
Products
Product Category
stains (1)
Brand
Sigma-Aldrich (2)
Formula Weight
pH Value
Physical Form
Chemical Composition
Quality Segment
Application
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
861324
p
-Rosolic acid,
Dye content 84 %
Expand
View Pricing
27-0620
p
-Rosolic acid,
JIS special grade
Expand
View Pricing