
Prohexadione-Calcium

Synonym(s):
3,5-Dioxo-4-propionyl-cyclohexanecarboxylic acid calcium salt
Empirical Formula (Hill Notation):
C10H10CaO5
CAS Number:
Molecular Weight:
250.26
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing