Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Probumin
®
Bovine Serum Albumin Vaccine Grade
CAS Number:
9048-46-8
Products
Product Category
albumins (4)
Brand
Sigma-Aldrich (4)
Biological Source
Physical Form
Tissue
Quality Segment
Geographic Origin
Package Size
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
840653
Probumin
®
Bovine Serum Albumin Vaccine Grade,
≥95%, USA origin, moisture (loss on drying): 0-10% ; heavy metals : 0-50 μg/g , microbiological culture, Adjuvant for veterinary use
Expand
View Pricing
840655
Probumin
®
Bovine Serum Albumin Vaccine Grade,
≥95%, USA origin, moisture (loss on drying): 0-10% ; heavy metals : 0-50 μg/g , microbiological culture, Adjuvant for veterinary use
Expand
View Pricing
840651
Probumin
®
Bovine Serum Albumin Vaccine Grade,
≥95%, USA origin, moisture (loss on drying): 0-10% ; heavy metals : 0-50 μg/g
Expand
View Pricing
840656
Probumin
®
Bovine Serum Albumin Vaccine Grade,
≥95%, USA origin, moisture (loss on drying): 0-10% ; heavy metals : 0-50 μg/g , microbiological culture, Adjuvant for veterinary use
Expand
View Pricing