Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Probumin
®
Bovine Serum Albumin Biotech Grade
CAS Number:
9048-46-8
Products
Product Category
albumins (3)
Brand
Sigma-Aldrich (3)
Biological Source
Physical Form
Tissue
Quality Segment
Geographic Origin
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
820472
Probumin
®
Bovine Serum Albumin Biotech Grade,
pkg of 10 g, ≥98%, USA origin, Tested for IgG, cell culture | mammalian, Binding, transport studies
Expand
View Pricing
820473
Probumin
®
Bovine Serum Albumin Biotech Grade,
pkg of 100 g, ≥98%, USA origin, Tested for IgG, cell culture | mammalian, Binding, transport studies
Expand
View Pricing
820476
Probumin
®
Bovine Serum Albumin Biotech Grade,
pkg of 5 kg, ≥98%, USA origin, Tested for IgG, cell culture | mammalian, Binding, transport studies
Expand
View Pricing