
Pregabalin

Synonym(s):
Pregabalin, (S)-3-(Aminomethyl)-5-methylhexanoic acid, CI-1008, PD-144723
Empirical Formula (Hill Notation):
C8H17NO2
CAS Number:
Molecular Weight:
159.23
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3