Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Potassium
tert
-butoxide
Synonym(s):
Potassium
tert
-butylate, Potassium
t
-butoxide
Linear Formula:
(CH
3
)
3
COK
CAS Number:
865-47-4
Molecular Weight:
112.21
EC Number:
212-740-3
Products
Product Category
organic bases (1)
Brand
Sigma-Aldrich (2)
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Core
Special Grade
Sublimed grade (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
156671
Potassium
tert
-butoxide,
reagent grade, ≥98%
Expand
View Pricing
659878
Potassium
tert
-butoxide,
sublimed grade, 99.99% trace metals basis
Expand
View Pricing