
Potassium hydrogen carbonate

Synonym(s):
Potassium hydrogen carbonate
Empirical Formula (Hill Notation):
CHKO3
CAS Number:
Molecular Weight:
100.12
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Agency
Quality Segment
Application
Showing 1-4 of 4