Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Potassium hexachloroiridate(IV)
Synonym(s):
Iridium potassium chloride
Linear Formula:
K
2
IrCl
6
CAS Number:
16920-56-2
Molecular Weight:
483.13
EC Number:
240-976-7
Products
Formula Weight
Melting Point (°C)
Physical Form
Purity
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
208981
Potassium hexachloroiridate(IV),
technical grade
Expand
View Pricing
450162
Potassium hexachloroiridate(IV),
99.99% trace metals basis
Expand
View Pricing