Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Potassium chlorate
Synonym(s):
Potassium chlorate
Linear Formula:
KClO
3
CAS Number:
3811-04-9
Molecular Weight:
122.55
EC Number:
223-289-7
Products
Product Category
salts (5)
Brand
Sigma-Aldrich (4)
Supelco (1)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Special Grade
ACS reagent (3)
Puriss (2)
Anhydrous (1)
Puriss P.A. (1)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
255572
Potassium chlorate,
ACS reagent, ≥99.0%
Expand
View Pricing
1.04944
Potassium chlorate,
for analysis EMSURE
®
ACS,Reag. Ph Eur
Expand
View Pricing
12634
Potassium chlorate,
puriss., 99-101%
Expand
View Pricing
799386
Potassium chlorate,
anhydrous, free-flowing, Redi-Dri
™
, ACS reagent, ≥99%
Expand
View Pricing
31247
Potassium chlorate,
puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Expand
View Pricing