Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Potassium acetate
Synonym(s):
Potassium acetate
Linear Formula:
CH
3
COOK
CAS Number:
127-08-2
Molecular Weight:
98.14
EC Number:
204-822-2
Products
Product Category
salts (7)
buffers (4)
aroma chemicals (1)
preservatives (1)
Brand
SAFC (1)
Sigma-Aldrich (11)
Supelco (1)
Feature
Formula Weight
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Special Grade
ACS reagent (2)
Halal (1)
Puriss (1)
pharmaceutical primary standard (1)
Filter Products
Showing 1-14 of 14
Compare
Product Number
Description
Pricing
236497
Potassium acetate,
ACS reagent, ≥99.0%
Expand
View Pricing
P1190
Potassium acetate,
for molecular biology, ≥99.0%
Expand
View Pricing
P1147
Potassium acetate,
ReagentPlus
®
, ≥99.0%
Expand
View Pricing
1.04820
Potassium acetate,
EMPROVE
®
ESSENTIAL, Ph. Eur., BP, JPE, E 261
Expand
View Pricing
791733
Potassium acetate,
anhydrous, free-flowing, Redi-Dri
™
, ACS reagent, ≥99.0%
Expand
View Pricing
W292001
Potassium acetate,
98%
Expand
View Pricing
60035
Potassium acetate,
BioUltra, for molecular biology, ≥99.0% (NT)
Expand
View Pricing
25059
Potassium acetate,
puriss., meets analytical specification of Ph. Eur., BP, E261, 99-101%
Expand
View Pricing
255785
Potassium acetate,
99.98% trace metals basis
Expand
View Pricing
P5708
Potassium acetate,
BioXtra, ≥99.0%
Expand
View Pricing
1.93714
Potassium acetate,
EMPLURA
®
Expand
View Pricing
1549807
Potassium acetate,
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing
P1222
Potassium acetate,
meets USP testing specifications
Expand
View Pricing
24-3920
Potassium acetate,
SAJ first grade, ≥95.0%
Expand
View Pricing