Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Porapak
™
Porous Polymer Adsorbent
Products
Physical Form
Quality Segment
Package Size
Technique
Filter Products
Showing 1-14 of 14
Compare
Product Number
Description
Pricing
20331
Porapak
™
Porous Polymer Adsorbent,
matrix Porapak Q, 80-100 mesh, bottle of 75 cc
Expand
View Pricing
20330-U
Porapak
™
Porous Polymer Adsorbent,
matrix Porapak Q, 50-80 mesh, bottle of 75 cc
Expand
View Pricing
20325
Porapak
™
Porous Polymer Adsorbent,
matrix Porapak N, 80-100 mesh, bottle of 75 cc
Expand
View Pricing
20332
Porapak
™
Porous Polymer Adsorbent,
matrix Porapak Q, 100-120 mesh, bottle of 75 cc
Expand
View Pricing
20324
Porapak
™
Porous Polymer Adsorbent,
matrix Porapak N, 50-80 mesh, bottle of 75 cc
Expand
View Pricing
20343
Porapak
™
Porous Polymer Adsorbent,
matrix Porapak QS, 80-100 mesh, bottle of 75 cc
Expand
View Pricing
20328
Porapak
™
Porous Polymer Adsorbent,
matrix Porapak P, 80-100 mesh, bottle of 75 cc
Expand
View Pricing
20334
Porapak
™
Porous Polymer Adsorbent,
matrix Porapak R, 80-100 mesh, bottle of 75 cc
Expand
View Pricing
20329
Porapak
™
Porous Polymer Adsorbent,
matrix Porapak P, 100-120 mesh, bottle of 75 cc
Expand
View Pricing
20342
Porapak
™
Porous Polymer Adsorbent,
matrix Porapak QS, 50-80 mesh, bottle of 75 cc
Expand
View Pricing
20333
Porapak
™
Porous Polymer Adsorbent,
matrix Porapak R, 50-80 mesh, bottle of 75 cc
Expand
View Pricing
20344
Porapak
™
Porous Polymer Adsorbent,
matrix Porapak QS, 100-120 mesh, bottle of 75 cc
Expand
View Pricing
20337
Porapak
™
Porous Polymer Adsorbent,
matrix Porapak S, 80-100 mesh, bottle of 75 cc
Expand
View Pricing
20338
Porapak
™
Porous Polymer Adsorbent,
matrix Porapak S, 100-120 mesh, bottle of 75 cc
Expand
View Pricing