Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Polypropylene
Linear Formula:
[CH
2
CH(CH
3
)]
n
CAS Number:
9003-07-0
Products
Brand
Sigma-Aldrich (4)
Melting Point (°C)
Physical Form
Quality Segment
Application
Polymer Composition
Polymer Type
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
428116
Polypropylene,
Isotactic, average M
w
~12,000, average M
n
~5,000
Expand
View Pricing
427888
Polypropylene,
Isotactic, average M
w
~250,000, average M
n
~67,000
Expand
View Pricing
428175
Polypropylene,
Amorphous
Expand
View Pricing
427861
Polypropylene,
Isotactic, average M
w
~340,000, average M
n
~97,000
Expand
View Pricing