Skip to Content
Merck
Poly[(isobutylene-alt-maleic acid, ammonium salt)-co-(isobutylene-alt-maleic anhydride)] average Mw ~60,000

Poly[(isobutylene-alt-maleic acid, ammonium salt)-co-(isobutylene-alt-maleic anhydride)]

Poly[(isobutylene-alt-maleic acid, ammonium salt)-co-(isobutylene-alt-maleic anhydride)] average Mw ~60,000
Synonym(s):
maleated copolymer, maleated polymer, maleic copolymer
Linear Formula:
(C4H2O3 · C4H8)m · xH3N
CAS Number:

Products

pH Value
Polymer Composition
Polymer Type