Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Polychlorodibenzo-p-dioxins (PCDDs) and polychlorodibenzofurans (PCDFs) solution
Products
Manufacturer
Agency
Format
Reference Material Type
Special Grade
certified reference material (10)
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
BCR614
Polychlorodibenzo-p-dioxins (PCDDs) and polychlorodibenzofurans (PCDFs) solution,
BCR
®
, certified reference material
Expand
View Pricing
BCR614S0
Polychlorodibenzo-p-dioxins (PCDDs) and polychlorodibenzofurans (PCDFs) solution,
BCR
®
, certified reference material, Solution S0
Expand
View Pricing
BCR614S3
Polychlorodibenzo-p-dioxins (PCDDs) and polychlorodibenzofurans (PCDFs) solution,
BCR
®
, certified reference material, Solution S3
Expand
View Pricing
BCR614S7
Polychlorodibenzo-p-dioxins (PCDDs) and polychlorodibenzofurans (PCDFs) solution,
BCR
®
, certified reference material, Solution S7
Expand
View Pricing
BCR614S6
Polychlorodibenzo-p-dioxins (PCDDs) and polychlorodibenzofurans (PCDFs) solution,
BCR
®
, certified reference material, Solution S6
Expand
View Pricing
BCR614S8
Polychlorodibenzo-p-dioxins (PCDDs) and polychlorodibenzofurans (PCDFs) solution,
BCR
®
, certified reference material, Solution S8
Expand
View Pricing
BCR614S2
Polychlorodibenzo-p-dioxins (PCDDs) and polychlorodibenzofurans (PCDFs) solution,
BCR
®
, certified reference material, Solution S2
Expand
View Pricing
BCR614S4
Polychlorodibenzo-p-dioxins (PCDDs) and polychlorodibenzofurans (PCDFs) solution,
BCR
®
, certified reference material, Solution S4
Expand
View Pricing
BCR614S5
Polychlorodibenzo-p-dioxins (PCDDs) and polychlorodibenzofurans (PCDFs) solution,
BCR
®
, certified reference material, Solution S5
Expand
View Pricing
BCR614S1
Polychlorodibenzo-p-dioxins (PCDDs) and polychlorodibenzofurans (PCDFs) solution,
BCR
®
, certified reference material, Solution S1
Expand
View Pricing