Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Poly(4-styrenesulfonic acid-
co
-maleic acid) sodium salt
Linear Formula:
[CH
2
CH(C
6
H
4
SO
3
R)]
x
[CH(CO
2
R)CH(CO
2
R)]
y
, R=H or Na
CAS Number:
68037-40-1
Products
Physical Form
Polymer Composition
Polymer Type
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
434558
Poly(4-styrenesulfonic acid-
co
-maleic acid) sodium salt,
average M
w
~20,000, powder
Expand
View Pricing
434566
Poly(4-styrenesulfonic acid-
co
-maleic acid) sodium salt,
average M
w
~20,000, powder
Expand
View Pricing