Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pipette tips for Beckman
®
FX Robotics System
Products
Product Category
pipette tips (17)
Manufacturer
Color
Sterility
Filter Products
Showing 1-17 of 17
Compare
Product Number
Description
Pricing
AXYFX20R
Pipette tips for Beckman
®
FX Robotics System,
racked, capacity 20 μL, pkg of 5x960tips/cs
Expand
View Pricing
AXYFX250RS
Pipette tips for Beckman
®
FX Robotics System,
racked, capacity 250 μL, pkg of 5x960tips/cs, clear, sterile
Expand
View Pricing
AXYFX250R
Pipette tips for Beckman
®
FX Robotics System,
racked, capacity 250 μL, pkg of 5x960tips/cs, clear
Expand
View Pricing
AXYFXF200RS
Pipette tips for Beckman
®
FX Robotics System,
capacity 200 μL, racked, filtered, clear, sterile
Expand
View Pricing
AXYFX50R
Pipette tips for Beckman
®
FX Robotics System,
racked, pkg of 5x960tips/cs, clear
Expand
View Pricing
AXYFX384RS
Pipette tips for Beckman
®
FX Robotics System,
capacity 30 μL, pkg of 5x3840tips/cs, clear, sterile
Expand
View Pricing
AXYFX384XLRS
Pipette tips for Beckman
®
FX Robotics System,
extra long, racked, capacity 50 μL, pkg of 5x3840tips/cs, clear, sterile
Expand
View Pricing
AXYFX50RS
Pipette tips for Beckman
®
FX Robotics System,
racked, capacity 50 μL, pkg of 5x960tips/cs, clear, sterile
Expand
View Pricing
AXYFX10R
Pipette tips for Beckman
®
FX Robotics System,
racked, capacity 10 μL, pkg of 5x960tips/cs
Expand
View Pricing
AXYFX38430FPR
Pipette tips for Beckman
®
FX Robotics System,
fine point, racked, capacity 30 μL, pkg of 5x3840tips/cs
Expand
View Pricing
AXYFX38430FPRS
Pipette tips for Beckman
®
FX Robotics System,
fine point, racked, capacity 30 μL, pkg of 5x3840tips/cs, sterile
Expand
View Pricing
AXYFXF180RPB
Pipette tips for Beckman
®
FX Robotics System,
capacity 165 μL, with paper band, racked, filtered, clear
Expand
View Pricing
AXYFX20RS
Pipette tips for Beckman
®
FX Robotics System,
racked, capacity 20 μL, pkg of 5x960tips/cs, sterile
Expand
View Pricing
AXYFX384R
Pipette tips for Beckman
®
FX Robotics System,
racked, capacity 30 μL, pkg of 5x3840tips/cs, clear
Expand
View Pricing
AXYFX255WBR
Pipette tips for Beckman
®
FX Robotics System,
wide bore, racked, capacity 250 μL, pkg of 5x960tips/cs, clear
Expand
View Pricing
AXYFX384XLR
Pipette tips for Beckman
®
FX Robotics System,
extra long, racked, capacity 50 μL, pkg of 5x3840tips/cs, clear
Expand
View Pricing
AXYFX38445FPR
Pipette tips for Beckman
®
FX Robotics System,
fine point, racked, capacity 45 μL, pkg of 5x3840tips/cs
Expand
View Pricing