
Phthalylsulfathiazole

Synonym(s):
N4-Phthalylsulfathiazole, 2-[[[4-[(2-Thiazolylamino)sulfonyl]phenylamino]carbonyl]benzoic acid
Empirical Formula (Hill Notation):
C17H13N3O5S2
CAS Number:
Molecular Weight:
403.43
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing