Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Phosphatase, Alkaline from
Escherichia coli
Synonym(s):
Orthophosphoric-monoester phosphohydrolase (alkaline optimum)
CAS Number:
9001-78-9
EC Number:
232-631-4
Products
Product Category
enzymes (3)
Brand
Sigma-Aldrich (3)
Biological Source
Physical Form
Chemical Composition
Quality Segment
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
P5931
Phosphatase, Alkaline from
Escherichia coli
,
lyophilized powder, 30-60 units/mg protein (in glycine buffer)
Expand
View Pricing
P4252
Phosphatase, Alkaline from
Escherichia coli
,
ammonium sulfate suspension, 30-90 units/mg protein (modified Warburg-Christian, in glycine buffer)
Expand
View Pricing
P4069
Phosphatase, Alkaline from
Escherichia coli
,
buffered aqueous glycerol solution, 20-50 units/mg protein (in glycine buffer)
Expand
View Pricing