Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Petroleum ether
CAS Number:
8032-32-4
EC Number:
232-453-7
Products
Product Category
solvents (6)
Brand
Sigma-Aldrich (7)
Boiling Point (°C)
Physical Form
Purity
Quality Segment
Special Grade
ACS reagent (3)
Anhydrous (1)
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
184519
Petroleum ether,
ACS reagent
Expand
View Pricing
320447
Petroleum ether,
ACS reagent
Expand
View Pricing
261734
Petroleum ether,
spectrophotometric grade
Expand
View Pricing
673838
Petroleum ether,
ACS reagent
Expand
View Pricing
300314
Petroleum ether,
anhydrous
Expand
View Pricing
24-1190
Petroleum ether,
SAJ first grade, ≥90.0%
Expand
View Pricing
24-1180
Petroleum ether,
JIS special grade, ≥90.0%
Expand
View Pricing