Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Osmium tetroxide solution
Linear Formula:
OsO
4
CAS Number:
20816-12-0
Molecular Weight:
254.23
Products
Product Category
fixatives (2)
salts (2)
Brand
Sigma-Aldrich (4)
Color
Formula Weight
Physical Form
Application
Reagent Type
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
251755
Osmium tetroxide solution,
4 wt. % in H
2
O
Expand
View Pricing
75632
Osmium tetroxide solution,
suitable for electron microscopy, 4% in H
2
O
Expand
View Pricing
208868
Osmium tetroxide solution,
2.5 wt. % in
tert
-butanol
Expand
View Pricing
75633
Osmium tetroxide solution,
suitable for electron microscopy, 2% in H
2
O
Expand
View Pricing