ORBO™ 23 2-HMP on Amberlite™ XAD®-2 (20/40), 120/60 mg
Synonym(s):
ORBO™ 2-HMP on Supelpak™ 20N Tube
Products
Brand
Product Line
Agency
Chemical Composition
OD (mm)
Quality Segment
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing