Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
OneRack
®
Test Tube Rack
Products
Product Category
racks (50)
Color
Manufacturer
Diameter (mm)
Wells
Package Size
Filter Products
Showing 1-20 of 50
Page 1 of 3
Page 1 of 3
Compare
Product Number
Description
Pricing
HS27515B
OneRack
®
Test Tube Rack,
Full, polyoxymethylene (fiberglass reinforced), size 24 wells, well diam. 30 mm, blue
Expand
View Pricing
HS27505B
OneRack
®
Test Tube Rack,
Half, polyoxymethylene (fiberglass reinforced), size 9 wells, well diam. 30 mm, blue
Expand
View Pricing
HS27551B
OneRack
®
Test Tube Rack,
Full, polypropylene, size 72 wells, well diam. 13 mm, blue
Expand
View Pricing
HS27515A
OneRack
®
Test Tube Rack,
Full, polyoxymethylene (fiberglass reinforced), size 24 wells, well diam. 30 mm, white
Expand
View Pricing
HS27504C
OneRack
®
Test Tube Rack,
Half, polyoxymethylene (fiberglass reinforced), size 16 wells, well diam. 25 mm, green
Expand
View Pricing
HS27552B
OneRack
®
Test Tube Rack,
Full, polypropylene, size 72 wells, well diam. 16 mm, blue
Expand
View Pricing
HS27513A
OneRack
®
Test Tube Rack,
Full, polyoxymethylene (fiberglass reinforced), size 40 wells, well diam. 20 mm, white
Expand
View Pricing
HS120207
OneRack
®
Test Tube Rack,
Full, polyoxymethylene (fiberglass reinforced), size 24 wells, well diam. 30 mm, yellow
Expand
View Pricing
HS27552D
OneRack
®
Test Tube Rack,
Full, polypropylene, size 72 wells, well diam. 16 mm, red
Expand
View Pricing
HS27513B
OneRack
®
Test Tube Rack,
Full, polyoxymethylene (fiberglass reinforced), size 40 wells, well diam. 20 mm, blue
Expand
View Pricing
HS27552A
OneRack
®
Test Tube Rack,
Full, polypropylene, size 72 wells, well diam. 16 mm, white
Expand
View Pricing
HS27502C
OneRack
®
Test Tube Rack,
Half, polyoxymethylene (fiberglass reinforced), size 36 wells, well diam. 16 mm, green
Expand
View Pricing
HS27514D
OneRack
®
Test Tube Rack,
Full, polyoxymethylene (fiberglass reinforced), size 40 wells, well diam. 25 mm, red
Expand
View Pricing
HS27505C
OneRack
®
Test Tube Rack,
Half, polyoxymethylene (fiberglass reinforced), size 9 wells, well diam. 30 mm, green
Expand
View Pricing
HS27505D
OneRack
®
Test Tube Rack,
Half, polyoxymethylene (fiberglass reinforced), size 9 wells, well diam. 30 mm, red
Expand
View Pricing
HS27515D
OneRack
®
Test Tube Rack,
Full, polyoxymethylene (fiberglass reinforced), size 24 wells, well diam. 30 mm, red
Expand
View Pricing
HS27551D
OneRack
®
Test Tube Rack,
Full, polypropylene, size 72 wells, well diam. 13 mm, red
Expand
View Pricing
HS27551A
OneRack
®
Test Tube Rack,
Full, polypropylene, size 72 wells, well diam. 13 mm, white
Expand
View Pricing
HS27504A
OneRack
®
Test Tube Rack,
Half, polyoxymethylene (fiberglass reinforced), size 16 wells, well diam. 25 mm, white
Expand
View Pricing
HS27511A
OneRack
®
Test Tube Rack,
Full, polyoxymethylene (fiberglass reinforced), size 72 wells, well diam. 13 mm, white
Expand
View Pricing
Page 1 of 3
Page 1 of 3