Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Oncostatin M human
Synonym(s):
OSM
CAS Number:
106956-32-5
EC Number:
200-835-2
Products
Product Category
growth factors (2)
peptides (1)
Brand
Sigma-Aldrich (2)
Biological Source
Color
Gene Alias
Physical Form
Product Line
Purity
Recombinant Host
Quality Segment
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
H6541
Oncostatin M human,
OSM, recombinant, expressed in HEK 293 cells, HumanKine
®
, suitable for cell culture
Expand
View Pricing
O9635
Oncostatin M human,
BioReagent, ≥97% (SDS-PAGE), recombinant, expressed in
E. coli
, lyophilized powder, suitable for cell culture
Expand
View Pricing