
Oleylamine

Synonym(s):
cis-1-Amino-9-octadecene
Linear Formula:
CH3(CH2)7CH=CH(CH2)7CH2NH2
CAS Number:
Molecular Weight:
267.49
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Chemical Composition
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing