Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ohaus
®
pH Electrode
Products
Manufacturer
Package Size
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
OH83033966
Ohaus
®
pH Electrode,
ST210
Expand
View Pricing
OH83033965
Ohaus
®
pH Electrode,
ST310
Expand
View Pricing
OH30087569
Ohaus
®
pH Electrode,
STMICRO8
Expand
View Pricing
OH83033967
Ohaus
®
pH Electrode,
ST320
Expand
View Pricing
OH83033968
Ohaus
®
pH Electrode,
ST230
Expand
View Pricing
OH30087566
Ohaus
®
pH Electrode,
STMICRO5
Expand
View Pricing
OH83033969
Ohaus
®
pH Electrode,
STPURE
Expand
View Pricing
OH30129354
Ohaus
®
pH Electrode,
ST350
Expand
View Pricing
OH30393265
Ohaus
®
pH Electrode,
ST272
Expand
View Pricing
OH30129357
Ohaus
®
pH Electrode,
ST260
Expand
View Pricing
OH30240974
Ohaus
®
pH Electrode,
ST270
Expand
View Pricing
OH30129470
Ohaus
®
pH Electrode,
STSURF
Expand
View Pricing