Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nunc
®
DeepWell
™
96 well plates
Products
Product Category
plates (3)
Color
Manufacturer
Binding Type
Capacity (mL)
Wells
Working Volume (ml)
Color
Package Size
Pore size (µm)
Sterility
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
Z688681
Nunc
®
DeepWell
™
96 well plates,
20 μm pore size pore size, glass fiber membrane on top of PET frit non-treated, non-sterile, clear, polypropylene, case of 50
Expand
View Pricing
Z688673
Nunc
®
DeepWell
™
96 well plates,
20 μm pore size pore size, glass fiber (binding) membrane on top of PET frit, non-treated, non-sterile, clear, polypropylene, case of 50
Expand
View Pricing
Z688703
Nunc
®
DeepWell
™
96 well plates,
20 μm pore size pore size, glass fiber membrane non-treated, non-sterile, clear, polypropylene, case of 50
Expand
View Pricing