Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
N
,
N
-Diisopropylethylamine
Synonym(s):
N
-Ethyldiisopropylamine, DIPEA, Ethyldiisopropylamine, ‘Hünig’s base’
Linear Formula:
[(CH
3
)
2
CH]
2
NC
2
H
5
CAS Number:
7087-68-5
Molecular Weight:
129.24
EC Number:
230-392-0
Products
Product Category
amines (1)
solvents (1)
Brand
Sigma-Aldrich (1)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Special Grade
Biotechnology (1)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
496219
N
,
N
-Diisopropylethylamine,
99.5%, biotech. grade
Expand
View Pricing
D125806
N
,
N
-Diisopropylethylamine,
ReagentPlus
®
, ≥99%
Expand
View Pricing
387649
N
,
N
-Diisopropylethylamine,
purified by redistillation, 99.5%
Expand
View Pricing
900690
N
,
N
-Diisopropylethylamine,
purified by redistillation, ZerO
2
®
, 99.5%
Expand
View Pricing