
Nitrofurfural diacetate

Synonym(s):
5-Nitro-2-furaldehyde diacetate, 5-Nitrofurfurylidene diacetate
Empirical Formula (Hill Notation):
C9H9NO7
CAS Number:
Molecular Weight:
243.17
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3