Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nimonic
®
alloy 90, Ni53Cr20Co18Ti2.5Al1.5Fe1.5
Products
Manufacturer
Physical Form
Diameter (mm)
Length (mm)
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
GF90094800
Nimonic
®
alloy 90, Ni53Cr20Co18Ti2.5Al1.5Fe1.5,
rod, 200mm x 20mm diameter
Expand
View Pricing
GF89613276
Nimonic
®
alloy 90, Ni53Cr20Co18Ti2.5Al1.5Fe1.5,
rod, 500mm x 12.7mm diameter
Expand
View Pricing
GF61256022
Nimonic
®
alloy 90, Ni53Cr20Co18Ti2.5Al1.5Fe1.5,
rod, 500mm x 22mm diameter
Expand
View Pricing
GF44921684
Nimonic
®
alloy 90, Ni53Cr20Co18Ti2.5Al1.5Fe1.5,
rod, 100mm x 22mm diameter
Expand
View Pricing
GF66578530
Nimonic
®
alloy 90, Ni53Cr20Co18Ti2.5Al1.5Fe1.5,
rod, 100mm x 12.7mm diameter
Expand
View Pricing
GF31885500
Nimonic
®
alloy 90, Ni53Cr20Co18Ti2.5Al1.5Fe1.5,
rod, 100mm x 20mm diameter
Expand
View Pricing
GF46411017
Nimonic
®
alloy 90, Ni53Cr20Co18Ti2.5Al1.5Fe1.5,
rod, 200mm x 22mm diameter
Expand
View Pricing
GF22316903
Nimonic
®
alloy 90, Ni53Cr20Co18Ti2.5Al1.5Fe1.5,
rod, 200mm x 12.7mm diameter
Expand
View Pricing
GF65932421
Nimonic
®
alloy 90, Ni53Cr20Co18Ti2.5Al1.5Fe1.5,
rod, 500mm x 20mm diameter
Expand
View Pricing