
Nimodipine Related Compound B

Synonym(s):
Bis(2-methoxyethyl) 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
Empirical Formula (Hill Notation):
C21H26N2O8
CAS Number:
Molecular Weight:
434.44
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2