Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nickel/Iron
Products
Manufacturer
Physical Form
Length (m)
Length (mm)
OD (mm)
Filter Products
Showing 1-20 of 25
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
GF89092941
Nickel/Iron,
powder, Ni81%/Fe19%, 45 max. part. size (micron), weight 50 g, condition atomized
Expand
View Pricing
GF07092682
Nickel/Iron,
powder, Ni 81%/Fe 19%, max. particle size 45 micron, weight 200 g, condition atomized
Expand
View Pricing
GF71847951
Nickel/Iron,
sheet, Ni52%/Fe48%, thickness 7.1 mm, size 108 × 108 mm
Expand
View Pricing
GF18067305
Nickel/Iron,
wire, Ni 90%/Fe 10%, 0.5 mm diameter, length 1 m, temper as drawn
Expand
View Pricing
GF68266996
Nickel/Iron,
rod, Ni80%/Fe20%, 10.0 mm diameter, length 100 mm, temper as drawn
Expand
View Pricing
GF17539140
Nickel/Iron,
rod, Ni52%/Fe 48%, 2 mm diameter, length 200 mm, temper annealed
Expand
View Pricing
GF41728989
Nickel/Iron,
tube, Ni52%/Fe 48%, outside diameter 9.55 mm, length 500 mm, wall thickness 0.275 mm
Expand
View Pricing
GF66697240
Nickel/Iron,
rod, 2 mm diameter, Ni52%/Fe48%, length 1000 mm, temper annealed
Expand
View Pricing
GF17794761
Nickel/Iron,
rod, Ni80%/Fe 20%, 5.0 mm diameter, length 100 mm, temper as drawn
Expand
View Pricing
GF21307935
Nickel/Iron,
wire, Ni 90%/Fe 10%, 0.5 mm diameter, length 0.2 m, temper as drawn
Expand
View Pricing
GF23554638
Nickel/Iron,
rod, Ni52%/Fe 48%, 3 mm diameter, length 500 mm
Expand
View Pricing
GF30925997
Nickel/Iron,
wire, Ni 90%/Fe 10%, 0.5 mm diameter, length 0.1 m, temper as drawn
Expand
View Pricing
GF97150552
Nickel/Iron,
rod, Ni52%/Fe48%, 3 mm diameter, length 200 mm
Expand
View Pricing
GF59124292
Nickel/Iron,
rod, Ni52%/Fe 48%, 3 mm diameter, length 100 mm
Expand
View Pricing
GF06936598
Nickel/Iron,
rod, Ni52%/Fe 48%, 2 mm diameter, length 100 mm, temper annealed
Expand
View Pricing
GF17167888
Nickel/Iron,
rod, Ni80%/Fe 20%, 10.0 mm diameter, length 500 mm, temper as drawn
Expand
View Pricing
GF43035690
Nickel/Iron,
tube, Ni52%/Fe 48%, outside diameter 9.55 mm, length 1000 mm, wall thickness 0.275 mm
Expand
View Pricing
GF52247189
Nickel/Iron,
tube, Ni52%/Fe 48%, outside diameter 9.55 mm, length 200 mm, wall thickness 0.275 mm
Expand
View Pricing
GF86895679
Nickel/Iron,
rod, 5.0 mm diameter, Ni80%/Fe20%, length 200 mm, temper as drawn
Expand
View Pricing
GF85408858
Nickel/Iron,
rod, Ni80%/Fe20%, length 500 mm, 5.0 mm diameter, temper as drawn
Expand
View Pricing
Page 1 of 2
Page 1 of 2